The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173689.6 | NP_775960.4 | 2 - 3 | 510 | 132 |
XM_011518556.2 | XP_011516858.1 | 2 - 3 | 510 | 132 | |
XM_011518557.2 | XP_011516859.1 | 2 - 3 | 512 | 132 | |
XM_011518558.2 | XP_011516860.1 | 3 - 4 | 872 | 132 | |
GenBank mRNA | AK123000.1 | - | 2 - 3 | 500 | 132 |
AY720432.1 | - | 2 - 3 | 416 | 132 |