The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317134.1 | NP_001304063.1 | 13 - 14 | 1581 | 73 |
NM_001317135.1 | NP_001304064.1 | 12 - 13 | 1412 | 73 | |
NM_001317136.1 | NP_001304065.1 | 12 - 13 | 1319 | 73 | |
NM_001317137.1 | NP_001304066.1 | 11 - 12 | 1328 | 73 | |
NM_001317138.1 | NP_001304067.1 | 13 - 14 | 1561 | 73 | |
NM_004567.3 | NP_004558.1 | 12 - 13 | 1433 | 73 | |
XM_011533829.2 | XP_011532131.1 | 12 - 13 | 1987 | 73 | |
XM_017006614.1 | XP_016862103.1 | 13 - 14 | 1471 | 73 | |
XM_017006615.1 | XP_016862104.1 | 12 - 13 | 1366 | 73 | |
XM_017006616.1 | XP_016862105.1 | 12 - 13 | 1279 | 73 | |
XM_017006617.1 | XP_016862106.1 | 12 - 13 | 1370 | 73 | |
GenBank mRNA | AF108765.1 | - | 12 - 13 | 1309 | 73 |
AK298732.1 | - | 12 - 13 | 1268 | 73 | |
AY707863.1 | - | 12 - 13 | 1379 | 73 | |
AY714243.1 | - | 13 - 14 | 1548 | 73 | |
AY756062.1 | - | 13 - 14 | 1537 | 73 | |
AY756063.1 | - | 13 - 14 | 1528 | 73 | |
AY756064.1 | - | 11 - 12 | 1295 | 73 | |
BC010269.1 | - | 12 - 13 | 1304 | 73 | |
D49818.1 | - | 12 - 13 | 1313 | 73 |