The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317134.1 | NP_001304063.1 | 3 - 4 | 511 | 62 |
NM_001317135.1 | NP_001304064.1 | 2 - 3 | 363 | 62 | |
NM_001317136.1 | NP_001304065.1 | 2 - 3 | 249 | 62 | |
NM_001317137.1 | NP_001304066.1 | 2 - 3 | 363 | 62 | |
NM_001317138.1 | NP_001304067.1 | 2 - 3 | 363 | 62 | |
NM_004567.3 | NP_004558.1 | 2 - 3 | 363 | 62 | |
XM_011533829.2 | XP_011532131.1 | 2 - 3 | 917 | 62 | |
XM_017006614.1 | XP_016862103.1 | 3 - 4 | 401 | 62 | |
XM_017006615.1 | XP_016862104.1 | 3 - 4 | 401 | 62 | |
XM_017006616.1 | XP_016862105.1 | 2 - 3 | 209 | 62 | |
XM_017006617.1 | XP_016862106.1 | 2 - 3 | 300 | 62 | |
GenBank mRNA | AF108765.1 | - | 2 - 3 | 239 | 62 |
AK298732.1 | - | 2 - 3 | 198 | 62 | |
AK299097.1 | - | 2 - 3 | 264 | 62 | |
AK312623.1 | - | 2 - 3 | 271 | 62 | |
AY707863.1 | - | 2 - 3 | 330 | 62 | |
AY714243.1 | - | 3 - 4 | 478 | 62 | |
AY756062.1 | - | 3 - 4 | 467 | 62 | |
AY756063.1 | - | 2 - 3 | 330 | 62 | |
AY756064.1 | - | 2 - 3 | 330 | 62 | |
BC010269.1 | - | 2 - 3 | 234 | 62 | |
DC423860.1 | - | 2 - 3 | 249 | 62 |