The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317134.1 | NP_001304063.1 | 7 - 8 | 806 | 72 |
NM_001317135.1 | NP_001304064.1 | 6 - 7 | 658 | 72 | |
NM_001317136.1 | NP_001304065.1 | 6 - 7 | 544 | 72 | |
NM_001317137.1 | NP_001304066.1 | 6 - 7 | 658 | 72 | |
NM_001317138.1 | NP_001304067.1 | 7 - 8 | 786 | 72 | |
NM_004567.3 | NP_004558.1 | 6 - 7 | 658 | 72 | |
XM_011533829.2 | XP_011532131.1 | 6 - 7 | 1212 | 72 | |
XM_017006614.1 | XP_016862103.1 | 7 - 8 | 696 | 72 | |
XM_017006615.1 | XP_016862104.1 | 7 - 8 | 696 | 72 | |
XM_017006616.1 | XP_016862105.1 | 6 - 7 | 504 | 72 | |
XM_017006617.1 | XP_016862106.1 | 6 - 7 | 595 | 72 | |
GenBank mRNA | AF108765.1 | - | 6 - 7 | 534 | 72 |
AK298732.1 | - | 6 - 7 | 493 | 72 | |
AK299097.1 | - | 5 - 6 | 492 | 72 | |
AK312623.1 | - | 6 - 7 | 566 | 72 | |
AY707863.1 | - | 6 - 7 | 625 | 72 | |
AY714243.1 | - | 7 - 8 | 773 | 72 | |
AY756062.1 | - | 7 - 8 | 762 | 72 | |
AY756063.1 | - | 7 - 8 | 753 | 72 | |
AY756064.1 | - | 6 - 7 | 625 | 72 | |
BC010269.1 | - | 6 - 7 | 529 | 72 | |
D49818.1 | - | 6 - 7 | 538 | 72 | |
DC423860.1 | - | 6 - 7 | 544 | 72 |