The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001276293.1 | NP_001263222.1 | 6 - 7 | 806 | 114 |
NM_001276294.1 | NP_001263223.1 | 6 - 7 | 806 | 114 | |
NM_007184.3 | NP_009115.2 | 6 - 7 | 806 | 114 | |
XM_005264839.2 | XP_005264896.1 | 5 - 6 | 579 | 114 | |
GenBank mRNA | AB023192.1 | - | 7 - 8 | 744 | 114 |
AF082516.1 | - | 6 - 7 | 698 | 114 | |
AK001505.1 | - | 4 - 5 | 465 | 114 | |
AK027428.1 | - | 6 - 7 | 684 | 114 | |
AK291398.1 | - | 6 - 7 | 710 | 114 | |
AK315001.1 | - | 6 - 7 | 710 | 114 | |
BC038102.2 | - | 6 - 7 | 710 | 114 | |
BC054494.1 | - | 6 - 7 | 704 | 114 | |
BC056900.1 | - | 6 - 7 | 676 | 114 |