The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003041.3 | NP_003032.1 | 10 - 11 | 1302 | 104 |
XM_006721072.3 | XP_006721135.2 | 10 - 11 | 1885 | 104 | |
GenBank mRNA | M95549.1 | - | 10 - 11 | 1303 | 104 |