Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTPRC
Entrez Gene ID
5788
Gene Name
protein tyrosine phosphatase, receptor type C
Gene Aliases
B220, CD45, CD45R, GP180, L-CA, LCA, LY5, T200
Chromosome Location
Chr.1: 198638968 - 198757476 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002838.4 NP_002829.3 7 - 8 869 151
NM_080921.3 NP_563578.2 4 - 5 386 151
XM_006711472.3 XP_006711535.1 6 - 7 686 151
XM_006711473.3 XP_006711536.1 6 - 7 614 151
XM_006711474.3 XP_006711537.1 5 - 6 470 151
GenBank mRNA AK292131.1 - 4 - 5 259 151
AK299986.1 - 6 - 7 544 151
AK303539.1 - 7 - 8 742 151
AK307635.1 - 6 - 7 603 151
AK308217.1 - 5 - 6 476 151
BC014239.2 - 6 - 7 589 151
DA670254.1 - 4 - 5 289 151
DA819314.1 - 4 - 5 386 151
Y00062.1 - 5 - 6 313 151
Y00638.1 - 7 - 8 736 151

View Full Product Details