The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012112.4 | NP_036244.2 | 11 - 12 | 1896 | 59 |
XM_011528697.2 | XP_011526999.1 | 11 - 12 | 1454 | 59 | |
XM_011528699.2 | XP_011527001.1 | 10 - 11 | 1642 | 59 | |
XM_011528700.2 | XP_011527002.1 | 10 - 11 | 1375 | 59 | |
GenBank mRNA | AB024704.1 | - | 11 - 12 | 1669 | 59 |
AB027467.1 | - | 10 - 11 | 1305 | 59 | |
AF098158.1 | - | 11 - 12 | 1689 | 59 | |
AF146731.1 | - | 10 - 11 | 1302 | 59 | |
AF244547.1 | - | 8 - 9 | 1076 | 59 | |
AF287265.1 | - | 12 - 13 | 1770 | 59 | |
AK001391.1 | - | 7 - 8 | 979 | 59 | |
AK312549.1 | - | 11 - 12 | 1674 | 59 | |
AL117534.1 | - | 9 - 10 | 1140 | 59 | |
AY734680.1 | - | 9 - 10 | 1198 | 59 | |
BC004136.1 | - | 11 - 12 | 1626 | 59 | |
BC020207.1 | - | 11 - 12 | 1655 | 59 |