The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_206963.1 | NP_996846.1 | 4 - 5 | 744 | 121 |
XM_005247686.4 | XP_005247743.3 | 4 - 5 | 1160 | 121 | |
GenBank mRNA | AK130079.1 | - | 4 - 5 | 715 | 121 |
BC029640.1 | - | 4 - 5 | 680 | 121 |