Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
STAMBP
Entrez Gene ID
10617
Gene Name
STAM binding protein
Gene Aliases
AMSH, MICCAP
Chromosome Location
Chr.2: 73828911 - 73873656 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017003175.1 XP_016858664.1 1 - 2 32 74
XM_017003179.1 XP_016858668.1 1 - 2 33 74

View Full Product Details