Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DPH7
Entrez Gene ID
92715
Gene Name
diphthamide biosynthesis 7
Gene Aliases
C9orf112, RRT2, WDR85
Chromosome Location
Chr.9: 137554899 - 137578935 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138778.3 NP_620133.1 7 - 8 933 78
XM_005266123.2 XP_005266180.1 8 - 9 1391 78
XM_005266126.3 XP_005266183.1 6 - 7 1164 78
XM_005266128.2 XP_005266185.1 8 - 9 1017 78
XM_005266129.2 XP_005266186.1 8 - 9 1020 78
XM_005266130.2 XP_005266187.1 6 - 7 788 78
XM_011519189.1 XP_011517491.1 7 - 8 1034 78
XM_011519190.2 XP_011517492.1 7 - 8 922 78
XM_011519191.2 XP_011517493.1 7 - 8 922 78
XM_011519192.2 XP_011517494.1 7 - 8 922 78
XM_011519193.2 XP_011517495.1 7 - 8 922 78
XM_011519194.1 XP_011517496.1 7 - 8 922 78
XM_011519199.2 XP_011517501.1 7 - 8 879 78
XM_011519200.1 XP_011517502.1 7 - 8 882 78
XM_017015291.1 XP_016870780.1 8 - 9 1226 78
XM_017015292.1 XP_016870781.1 7 - 8 1296 78
XM_017015293.1 XP_016870782.1 7 - 8 1253 78
XM_017015294.1 XP_016870783.1 9 - 10 1206 78
XM_017015295.1 XP_016870784.1 7 - 8 1293 78
XM_017015296.1 XP_016870785.1 7 - 8 1132 78
XM_017015297.1 XP_016870786.1 9 - 10 1203 78
XM_017015298.1 XP_016870787.1 8 - 9 1486 78
XM_017015299.1 XP_016870788.1 8 - 9 743 78
XM_017015300.1 XP_016870789.1 8 - 9 1483 78
XM_017015301.1 XP_016870790.1 6 - 7 1156 78
XM_017015302.1 XP_016870791.1 6 - 7 2210 78
XM_017015303.1 XP_016870792.1 7 - 8 1391 78
XM_017015304.1 XP_016870793.1 6 - 7 856 78
GenBank mRNA AK075115.1 - 7 - 8 934 78
AK310226.1 - 3 - 4 227 78
BC003123.1 - 7 - 8 872 78
BC017335.2 - 7 - 8 892 78

View Full Product Details