Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LCN1
Entrez Gene ID
3933
Gene Name
lipocalin 1
Gene Aliases
PMFA, TLC, TP, VEGP
Chromosome Location
Chr.9: 135521296 - 135526540 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252617.1 NP_001239546.1 1 - 2 150 99
NM_001252618.1 NP_001239547.1 1 - 2 150 99
NM_001252619.1 NP_001239548.1 1 - 2 150 99
NM_002297.3 NP_002288.1 1 - 2 150 99
XM_011518675.2 XP_011516977.1 1 - 2 292 99
GenBank mRNA BC065721.1 - 1 - 2 152 99
BC074925.2 - 1 - 2 114 99
BC074926.2 - 1 - 2 114 99
CD721968.1 - 1 - 2 130 99
CK429694.1 - 1 - 2 127 99
CK430667.1 - 1 - 2 138 99
X62418.1 - 1 - 2 112 99

View Full Product Details