Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLA2G6
Entrez Gene ID
8398
Gene Name
phospholipase A2 group VI
Gene Aliases
CaI-PLA2, GVI, INAD1, IPLA2-VIA, NBIA2, NBIA2A, NBIA2B, PARK14, PLA2, PNPLA9, iPLA2, iPLA2beta
Chromosome Location
Chr.22: 38111495 - 38192099 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004426.1 NP_001004426.1 5 - 6 933 62
NM_001199562.1 NP_001186491.1 5 - 6 888 62
NM_003560.2 NP_003551.2 5 - 6 933 62
XM_005261764.2 XP_005261821.1 5 - 6 908 62
XM_005261765.1 XP_005261822.1 5 - 6 905 62
XM_005261766.1 XP_005261823.1 5 - 6 1000 62
XM_006724332.3 XP_006724395.1 5 - 6 909 62
XM_011530423.1 XP_011528725.1 6 - 7 1020 62
XM_011530424.1 XP_011528726.1 7 - 8 1144 62
XM_011530425.1 XP_011528727.1 5 - 6 844 62
XM_011530426.2 XP_011528728.1 5 - 6 908 62
XM_017028981.1 XP_016884470.1 5 - 6 1000 62
XM_017028982.1 XP_016884471.1 5 - 6 905 62
XM_017028983.1 XP_016884472.1 7 - 8 1144 62
XM_017028984.1 XP_016884473.1 6 - 7 1020 62
XM_017028986.1 XP_016884475.1 5 - 6 908 62
XM_017028987.1 XP_016884476.1 5 - 6 908 62
XM_017028988.1 XP_016884477.1 5 - 6 908 62
GenBank mRNA AF064594.1 - 5 - 6 933 62
AF102988.1 - 4 - 5 62
AF102989.1 - 4 - 5 62
AK001290.1 - - 222 62
AK290825.1 - 5 - 6 890 62
AK291212.1 - 5 - 6 919 62
AK308308.1 - 4 - 5 739 62
AL080187.1 - 5 - 6 933 62
BC036742.1 - 5 - 6 927 62
BC051904.1 - 5 - 6 869 62
CR456543.1 - 5 - 6 861 62

View Full Product Details