The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128928.1 | NP_001122400.1 | 4 - 5 | 964 | 132 |
NM_002194.3 | NP_002185.1 | 3 - 4 | 820 | 132 | |
XM_005246532.1 | XP_005246589.1 | 3 - 4 | 601 | 132 | |
GenBank mRNA | AB451298.1 | - | 2 - 3 | 264 | 132 |
AK093560.1 | - | 5 - 6 | 922 | 132 | |
BC015496.1 | - | 2 - 3 | 330 | 132 | |
BC106006.1 | - | 3 - 4 | 364 | 132 | |
CR456840.1 | - | 2 - 3 | 264 | 132 | |
CR541707.1 | - | 2 - 3 | 264 | 132 | |
KJ535023.1 | - | 2 - 3 | 264 | 132 | |
KU178146.1 | - | 2 - 3 | 264 | 132 | |
L08488.1 | - | 3 - 4 | 590 | 132 |