The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022437.2 | NP_071882.1 | 1 - 2 | 81 | |
XM_005264483.3 | XP_005264540.1 | 1 - 2 | 81 | ||
GenBank mRNA | AF320294.1 | - | 1 - 2 | 81 | |
AF324494.1 | - | 1 - 2 | 81 | ||
BC113657.1 | - | 1 - 2 | 99 | 81 | |
BC143564.1 | - | 1 - 2 | 99 | 81 |