The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001010982.4 | NP_001010982.2 | 4 - 5 | 360 | 74 |
NM_001145526.2 | NP_001138998.1 | 4 - 5 | 360 | 74 | |
XM_011524326.2 | XP_011522628.1 | 3 - 4 | 292 | 74 | |
XM_011524327.2 | XP_011522629.1 | 5 - 6 | 528 | 74 | |
XM_011524328.2 | XP_011522630.1 | 5 - 6 | 500 | 74 | |
XM_011524329.2 | XP_011522631.1 | 5 - 6 | 542 | 74 | |
XM_011524330.2 | XP_011522632.1 | 5 - 6 | 546 | 74 | |
XM_011524333.2 | XP_011522635.1 | 4 - 5 | 319 | 74 | |
XM_011524336.2 | XP_011522638.1 | 4 - 5 | 319 | 74 | |
GenBank mRNA | BC132824.1 | - | 4 - 5 | 360 | 74 |
BX648442.1 | - | 4 - 5 | 316 | 74 |