The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007202.3 | NP_009133.2 | 10 - 11 | 1812 | 99 |
XM_017024071.1 | XP_016879560.1 | 10 - 11 | 1807 | 99 | |
GenBank mRNA | AF037439.1 | - | 10 - 11 | 1772 | 99 |
AK308297.1 | - | 10 - 11 | 1659 | 99 | |
AK312438.1 | - | 10 - 11 | 1765 | 99 | |
BC017055.1 | - | 10 - 11 | 1768 | 99 | |
KU178546.1 | - | 10 - 11 | 1635 | 99 | |
KU178547.1 | - | 10 - 11 | 1606 | 99 | |
KU178548.1 | - | 9 - 10 | 1452 | 99 | |
KU178549.1 | - | 3 - 4 | 304 | 99 |