The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003934.1 | NP_003925.1 | 2 - 3 | 300 | 92 |
XM_005272232.1 | XP_005272289.1 | 2 - 3 | 306 | 92 | |
XM_006717313.2 | XP_006717376.1 | 3 - 4 | 1151 | 92 | |
XM_011519172.2 | XP_011517474.1 | 2 - 3 | 306 | 92 | |
XM_011519174.2 | XP_011517476.1 | 4 - 5 | 1396 | 92 | |
XM_017015269.1 | XP_016870758.1 | 3 - 4 | 1151 | 92 | |
XM_017015270.1 | XP_016870759.1 | 3 - 4 | 1151 | 92 | |
XM_017015271.1 | XP_016870760.1 | 4 - 5 | 1396 | 92 | |
XM_017015273.1 | XP_016870762.1 | 3 - 4 | 1151 | 92 | |
GenBank mRNA | AK295155.1 | - | 2 - 3 | 300 | 92 |
AL537190.3 | - | 2 - 3 | 271 | 92 | |
BC137338.1 | - | 2 - 3 | 297 | 92 | |
BC137340.1 | - | 2 - 3 | 297 | 92 | |
BI461266.1 | - | 2 - 3 | 304 | 91 |