The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003934.1 | NP_003925.1 | 4 - 5 | 384 | 138 |
XM_005272232.1 | XP_005272289.1 | 4 - 5 | 390 | 138 | |
XM_006717313.2 | XP_006717376.1 | 5 - 6 | 1235 | 138 | |
XM_011519172.2 | XP_011517474.1 | 4 - 5 | 390 | 138 | |
XM_011519173.2 | XP_011517475.1 | 2 - 3 | 642 | 138 | |
XM_011519174.2 | XP_011517476.1 | 6 - 7 | 1480 | 138 | |
XM_017015269.1 | XP_016870758.1 | 5 - 6 | 1235 | 138 | |
GenBank mRNA | AK295155.1 | - | 4 - 5 | 384 | 138 |
AL537190.3 | - | 4 - 5 | 355 | 138 | |
BC007874.2 | - | 5 - 6 | 454 | 138 | |
BC137338.1 | - | 4 - 5 | 381 | 138 | |
BC137340.1 | - | 4 - 5 | 381 | 138 | |
BI461266.1 | - | 4 - 5 | 388 | 138 | |
U69127.1 | - | 4 - 5 | 360 | 138 |