The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003119.3 | NP_003110.1 | 9 - 10 | 1355 | 92 |
XM_006721264.3 | XP_006721327.1 | 9 - 10 | 1364 | 92 | |
XM_017023597.1 | XP_016879086.1 | 9 - 10 | 1364 | 92 | |
XM_017023598.1 | XP_016879087.1 | 9 - 10 | 1364 | 92 | |
GenBank mRNA | BC035929.2 | - | 8 - 9 | 1138 | 92 |
BC036104.1 | - | 9 - 10 | 1340 | 92 | |
Y16610.1 | - | 9 - 10 | 1337 | 92 |