The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303103.1 | NP_001290032.1 | 12 - 13 | 2386 | 130 |
NM_005077.4 | NP_005068.2 | 12 - 13 | 2356 | 130 | |
XM_005252163.2 | XP_005252220.1 | 10 - 11 | 1048 | 130 | |
XM_011518951.2 | XP_011517253.1 | 12 - 13 | 1267 | 130 | |
XM_017015064.1 | XP_016870553.1 | 12 - 13 | 1297 | 130 | |
XM_017015066.1 | XP_016870555.1 | 10 - 11 | 1040 | 130 | |
GenBank mRNA | AK290860.1 | - | 12 - 13 | 1118 | 130 |
AK304565.1 | - | 10 - 11 | 957 | 130 | |
BC010100.2 | - | 12 - 13 | 1502 | 130 | |
BC015747.1 | - | 12 - 13 | 1212 | 130 | |
M99435.1 | - | 12 - 13 | 1086 | 130 |