Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- TLE1
- Entrez Gene ID
-
7088
- Gene Name
- transducin like enhancer of split 1
- Gene Aliases
-
ESG, ESG1, GRG1
- Chromosome Location
-
Chr.9: 81583683 - 81689535 on Build GRCh38
View Full Product Details