Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GSG1L
Entrez Gene ID
146395
Gene Name
GSG1 like
Gene Aliases
PRO19651
Chromosome Location
Chr.16: 27787529 - 28063509 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BI752847.1 - 3 - 4 309 65

View Full Product Details