Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GSG1L
Entrez Gene ID
146395
Gene Name
GSG1 like
Gene Aliases
PRO19651
Chromosome Location
Chr.16: 27787529 - 28063509 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109763.1 NP_001103233.1 4 - 5 744 78
NM_001323900.1 NP_001310829.1 4 - 5 744 78
NM_001323901.1 NP_001310830.1 3 - 4 591 78
NM_144675.2 NP_653276.1 3 - 4 360 78
XM_011545750.2 XP_011544052.1 4 - 5 738 78
GenBank mRNA AY302134.1 - 2 - 3 194 78
AY358206.1 - 3 - 4 583 78
BC016460.1 - 3 - 4 360 78
BI752847.1 - 5 - 6 567 78
CV024949.1 - 2 - 3 194 78

View Full Product Details