The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318204.1 | NP_001305133.1 | 2 - 3 | 364 | 107 |
NM_004560.3 | NP_004551.2 | 2 - 3 | 364 | 107 | |
XM_005252008.4 | XP_005252065.1 | 2 - 3 | 329 | 107 | |
XM_006717121.3 | XP_006717184.1 | 2 - 3 | 986 | 107 | |
XM_017014762.1 | XP_016870251.1 | 2 - 3 | 2061 | 107 | |
XM_017014763.1 | XP_016870252.1 | 2 - 3 | 240 | 107 | |
GenBank mRNA | AB209154.1 | - | 4 - 5 | 633 | 107 |
BC033697.1 | - | 2 - 3 | 351 | 107 | |
BC051273.1 | - | 2 - 3 | 176 | 107 | |
BC130522.1 | - | 2 - 3 | 252 | 107 | |
M97639.1 | - | 2 - 3 | 364 | 107 |