The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004560.3 | NP_004551.2 | 8 - 9 | 1586 | 68 |
XM_005252008.4 | XP_005252065.1 | 8 - 9 | 1551 | 68 | |
XM_005252009.3 | XP_005252066.1 | 2 - 3 | 292 | 68 | |
XM_006717121.3 | XP_006717184.1 | 8 - 9 | 2208 | 68 | |
XM_017014762.1 | XP_016870251.1 | 8 - 9 | 3283 | 68 | |
XM_017014763.1 | XP_016870252.1 | 8 - 9 | 1462 | 68 | |
GenBank mRNA | AB209154.1 | - | 10 - 11 | 1855 | 68 |
BC033697.1 | - | 8 - 9 | 1573 | 68 | |
BC130522.1 | - | 8 - 9 | 1474 | 68 | |
M97639.1 | - | 8 - 9 | 1586 | 68 |