The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003747.2 | NP_003738.2 | 14 - 15 | 2154 | 80 |
XM_006716263.3 | XP_006716326.1 | 14 - 15 | 2860 | 80 | |
XM_011543845.2 | XP_011542147.1 | 14 - 15 | 2860 | 80 | |
XM_011543846.2 | XP_011542148.1 | 14 - 15 | 2860 | 80 | |
GenBank mRNA | AB209339.1 | - | 13 - 14 | 1334 | 80 |
AF082556.1 | - | 14 - 15 | 2154 | 80 | |
AF082557.1 | - | 13 - 14 | 1450 | 80 | |
AK302534.1 | - | 14 - 15 | 1828 | 80 | |
BC098394.1 | - | 14 - 15 | 2177 | 80 |