The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001272072.1 | NP_001259001.1 | 3 - 4 | 1957 | 116 |
XM_017011944.1 | XP_016867433.1 | 5 - 6 | 2165 | 116 | |
XM_017011945.1 | XP_016867434.1 | 5 - 6 | 2195 | 116 | |
XM_017011946.1 | XP_016867435.1 | 3 - 4 | 2121 | 116 | |
GenBank mRNA | AK314806.1 | - | 3 - 4 | 1915 | 116 |
U11863.1 | - | 3 - 4 | 1932 | 116 |