The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134367.3 | NP_001127839.2 | 8 - 9 | 1285 | 117 |
NM_003043.5 | NP_003034.2 | 8 - 9 | 1289 | 117 | |
XM_006713307.2 | XP_006713370.1 | 7 - 8 | 1093 | 117 | |
XM_011534030.2 | XP_011532332.1 | 7 - 8 | 1046 | 117 | |
XM_011534031.2 | XP_011532333.1 | 9 - 10 | 1417 | 117 | |
GenBank mRNA | BC137128.1 | - | 8 - 9 | 1236 | 117 |
BC137129.1 | - | 8 - 9 | 1232 | 117 | |
U09220.1 | - | 6 - 7 | 964 | 117 | |
U16120.1 | - | 9 - 10 | 1340 | 117 | |
Z18956.1 | - | 6 - 7 | 983 | 117 |