The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000316.2 | NP_000307.1 | 5 - 6 | 516 | 74 |
NM_001184744.1 | NP_001171673.1 | 3 - 4 | 379 | 74 | |
XM_005265344.3 | XP_005265401.1 | 3 - 4 | 2313 | 74 | |
XM_011533967.2 | XP_011532269.1 | 5 - 6 | 611 | 74 | |
XM_011533968.2 | XP_011532270.1 | 4 - 5 | 458 | 74 | |
XM_017006932.1 | XP_016862421.1 | 4 - 5 | 563 | 74 | |
XM_017006933.1 | XP_016862422.1 | 4 - 5 | 665 | 74 | |
XM_017006934.1 | XP_016862423.1 | 5 - 6 | 581 | 74 | |
GenBank mRNA | AY449732.1 | - | 3 - 4 | 313 | 74 |
BC110388.1 | - | 3 - 4 | 379 | 74 | |
BC112221.1 | - | 5 - 6 | 516 | 74 | |
BC112247.1 | - | 5 - 6 | 516 | 74 | |
L04308.1 | - | 3 - 4 | 341 | 74 | |
U17418.1 | - | 5 - 6 | 531 | 74 | |
X68596.1 | - | 5 - 6 | 487 | 74 |