The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270398.1 | NP_001257327.1 | 2 - 3 | 467 | 67 |
NM_002821.4 | NP_002812.2 | 2 - 3 | 594 | 67 | |
NM_152880.3 | NP_690619.1 | 2 - 3 | 594 | 67 | |
NM_152881.3 | NP_690620.1 | 2 - 3 | 594 | 67 | |
NM_152882.3 | NP_690621.1 | 2 - 3 | 594 | 67 | |
NR_072997.1 | 2 - 3 | 594 | 67 | ||
NR_072998.1 | 2 - 3 | 594 | 67 | ||
XM_011514765.2 | XP_011513067.1 | 2 - 3 | 462 | 67 | |
XM_011514766.2 | XP_011513068.1 | 2 - 3 | 462 | 67 | |
GenBank mRNA | AB209350.1 | - | 2 - 3 | 493 | 67 |
AF531868.1 | - | 2 - 3 | 520 | 67 | |
AF531869.1 | - | 2 - 3 | 520 | 67 | |
AF531870.1 | - | 2 - 3 | 520 | 67 | |
AF531871.1 | - | 2 - 3 | 520 | 67 | |
AF531872.1 | - | 2 - 3 | 520 | 67 | |
AK291016.1 | - | 2 - 3 | 426 | 67 | |
AK292589.1 | - | 2 - 3 | 521 | 67 | |
AK296931.1 | - | 2 - 3 | 443 | 67 | |
AK296953.1 | - | 2 - 3 | 402 | 67 | |
BC046109.2 | - | 2 - 3 | 534 | 67 | |
BC071557.1 | - | 2 - 3 | 548 | 67 | |
BM811107.1 | - | - | 68 | ||
U33635.1 | - | 2 - 3 | 565 | 67 | |
U40271.2 | - | 2 - 3 | 520 | 67 |