The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321042.1 | NP_001307971.1 | 7 - 8 | 1203 | 104 |
NM_005539.4 | NP_005530.3 | 10 - 11 | 1150 | 104 | |
XM_017016204.1 | XP_016871693.1 | 9 - 10 | 804 | 104 | |
XM_017016205.1 | XP_016871694.1 | 10 - 11 | 1145 | 104 | |
GenBank mRNA | AF273055.1 | - | 10 - 11 | 1093 | 104 |
BC096280.2 | - | 11 - 12 | 894 | 104 | |
BC096281.2 | - | 10 - 11 | 813 | 104 | |
X77567.1 | - | 10 - 11 | 931 | 104 | |
Z31695.1 | - | 10 - 11 | 786 | 104 |