The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321042.1 | NP_001307971.1 | 4 - 5 | 896 | 102 |
NM_005539.4 | NP_005530.3 | 7 - 8 | 843 | 102 | |
XM_017016204.1 | XP_016871693.1 | 6 - 7 | 497 | 102 | |
XM_017016205.1 | XP_016871694.1 | 7 - 8 | 838 | 102 | |
GenBank mRNA | AF273055.1 | - | 7 - 8 | 786 | 102 |
BC052265.1 | - | 6 - 7 | 441 | 102 | |
BC062300.1 | - | 6 - 7 | 441 | 102 | |
BC096280.2 | - | 8 - 9 | 587 | 102 | |
BC096281.2 | - | 7 - 8 | 506 | 102 | |
X77567.1 | - | 7 - 8 | 624 | 102 | |
Z31695.1 | - | 7 - 8 | 101 |