The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001200049.2 | NP_001186978.2 | 3 - 4 | 404 | 68 |
GenBank mRNA | AK098820.1 | - | 3 - 4 | 379 | 68 |
BC044661.1 | - | 3 - 4 | 362 | 68 | |
BC146900.1 | - | 2 - 3 | 187 | 68 | |
BC146908.1 | - | 2 - 3 | 187 | 68 | |
BX376477.2 | - | 3 - 4 | 404 | 68 | |
EL734348.1 | - | 3 - 4 | 303 | 68 |