Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CFAP46
Entrez Gene ID
54777
Gene Name
cilia and flagella associated protein 46
Gene Aliases
C10orf123, C10orf124, C10orf92, C10orf93, TTC40, bA288G11.4, bA288G11.5, bB137A17.2, bB137A17.3
Chromosome Location
Chr.10: 132808392 - 132942585 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001200049.2 NP_001186978.2 4 - 5 477 88
GenBank mRNA AK098820.1 - 4 - 5 452 88
BC044661.1 - 4 - 5 435 88
BC146900.1 - 3 - 4 260 88
BC146908.1 - 3 - 4 260 88
BX376477.2 - 4 - 5 477 88
EL734348.1 - 4 - 5 376 88

View Full Product Details