The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001258313.1 | NP_001245242.1 | 1 - 2 | 475 | 84 |
XM_011511325.2 | XP_011509627.1 | 1 - 2 | 470 | 84 | |
XM_017004302.1 | XP_016859791.1 | 1 - 2 | 470 | 84 | |
GenBank mRNA | AF110235.1 | - | 1 - 2 | 449 | 84 |
AJ401610.1 | - | 1 - 2 | 57 | 84 | |
AL110263.1 | - | 1 - 2 | 449 | 84 | |
DB128091.1 | - | 1 - 2 | 475 | 84 |