Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ULK3
Entrez Gene ID
25989
Gene Name
unc-51 like kinase 3
Gene Aliases
-
Chromosome Location
Chr.15: 74836116 - 74843211 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099436.3 NP_001092906.3 3 - 4 475 67
NM_001284364.2 NP_001271293.2 3 - 4 475 67
NM_001284365.2 NP_001271294.1 3 - 4 398 67
NR_104307.2 3 - 4 475 67
NR_104308.2 3 - 4 475 67
NR_104309.2 3 - 4 475 67
XM_005254289.2 XP_005254346.1 4 - 5 490 67
XM_017022068.1 XP_016877557.1 4 - 5 524 67
XM_017022069.1 XP_016877558.1 4 - 5 528 67
XM_017022070.1 XP_016877559.1 4 - 5 541 67
XM_017022071.1 XP_016877560.1 4 - 5 528 67
XM_017022072.1 XP_016877561.1 3 - 4 704 67
GenBank mRNA AK293181.1 - 3 - 4 328 67
AK293650.1 - 2 - 3 493 67
AK294241.1 - 3 - 4 409 67
AK294245.1 - 3 - 4 405 67
AK294251.1 - 3 - 4 409 67
AK298879.1 - 3 - 4 332 67
AK299380.1 - 3 - 4 402 67
AK316102.1 - 3 - 4 409 67
AL117482.1 - 3 - 4 610 67
BC036117.1 - 3 - 4 446 67
BC048289.1 - 3 - 4 403 67
BC056423.1 - 3 - 4 393 67
BC157884.1 - 3 - 4 457 67
BI829133.1 - 2 - 3 559 67
BQ927030.1 - 3 - 4 363 67

View Full Product Details