Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCN1A
Entrez Gene ID
6323
Gene Name
sodium voltage-gated channel alpha subunit 1
Gene Aliases
EIEE6, FEB3, FEB3A, FHM3, GEFSP2, HBSCI, NAC1, Nav1.1, SCN1, SMEI
Chromosome Location
Chr.2: 165989160 - 166149216 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001165963.1 NP_001159435.1 7 - 8 1048 96
NM_001165964.1 NP_001159436.1 7 - 8 1048 96
NM_001202435.1 NP_001189364.1 9 - 10 1257 96
NM_006920.4 NP_008851.3 7 - 8 1048 96
XM_011511602.2 XP_011509904.1 8 - 9 1324 96
XM_011511604.2 XP_011509906.1 8 - 9 1217 96
XM_011511605.2 XP_011509907.1 9 - 10 1424 96
XM_011511606.2 XP_011509908.1 9 - 10 1416 96
XM_017004644.1 XP_016860133.1 10 - 11 1503 96
XM_017004645.1 XP_016860134.1 9 - 10 1424 96
XM_017004646.1 XP_016860135.1 8 - 9 1338 96
XM_017004647.1 XP_016860136.1 10 - 11 1510 96
XM_017004648.1 XP_016860137.1 9 - 10 1287 96
XM_017004649.1 XP_016860138.1 8 - 9 1249 96
XM_017004650.1 XP_016860139.1 8 - 9 1326 96
XM_017004651.1 XP_016860140.1 9 - 10 1279 96
XM_017004652.1 XP_016860141.1 8 - 9 1319 96
XM_017004653.1 XP_016860142.1 9 - 10 1429 96
XM_017004654.1 XP_016860143.1 9 - 10 1440 96
GenBank mRNA AB093548.1 - 7 - 8 1030 96
AB093549.1 - 7 - 8 1030 96
AB098335.1 - 7 - 8 1030 96
AF225985.1 - 7 - 8 1048 96

View Full Product Details