The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001935.3 | NP_001926.2 | 2 - 3 | 656 | 57 |
GenBank mRNA | AB451339.1 | - | 1 - 2 | 95 | 57 |
AB451488.1 | - | 1 - 2 | 95 | 57 | |
AY429531.1 | - | 1 - 2 | 66 | 57 | |
AY429532.1 | - | 1 - 2 | 86 | 57 | |
BC013329.2 | - | 1 - 2 | 90 | 57 | |
BC065265.1 | - | 2 - 3 | 656 | 57 | |
M74777.1 | - | 2 - 3 | 105 | 57 | |
M80536.1 | - | 1 - 2 | 95 | 57 | |
X60708.1 | - | 2 - 3 | 170 | 57 |