Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM26
Entrez Gene ID
7726
Gene Name
tripartite motif containing 26
Gene Aliases
AFP, RNF95, ZNF173
Chromosome Location
Chr.6: 30184453 - 30213494 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242783.1 NP_001229712.1 1 - 2 192 74
XM_006715180.2 XP_006715243.1 1 - 2 122 74
XM_017011263.1 XP_016866752.1 1 - 2 133 74
GenBank mRNA BC032297.1 - 1 - 2 50 74

View Full Product Details