The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001045556.2 | NP_001039021.1 | 4 - 5 | 991 | 116 |
NM_001045557.2 | NP_001039022.2 | 3 - 4 | 713 | 116 | |
NM_001282964.1 | NP_001269893.1 | 3 - 4 | 713 | 116 | |
NM_001282965.1 | NP_001269894.1 | 2 - 3 | 612 | 116 | |
NM_006748.3 | NP_006739.2 | 2 - 3 | 357 | 116 | |
XM_017013739.1 | XP_016869228.1 | 3 - 4 | 1184 | 116 | |
GenBank mRNA | AJ238591.1 | - | 3 - 4 | 358 | 116 |
AK297423.1 | - | 3 - 4 | 259 | 116 | |
AK297519.1 | - | 2 - 3 | 148 | 116 | |
AK309401.1 | - | 3 - 4 | 339 | 116 | |
AK310092.1 | - | 3 - 4 | 296 | 116 | |
AK310115.1 | - | 3 - 4 | 374 | 116 | |
AK311010.1 | - | 3 - 4 | 300 | 116 | |
AK312584.1 | - | 3 - 4 | 523 | 116 | |
BC007042.1 | - | 2 - 3 | 298 | 116 | |
BT019994.1 | - | 2 - 3 | 159 | 116 | |
BX648382.1 | - | 4 - 5 | 574 | 116 | |
CR536537.1 | - | 2 - 3 | 159 | 116 | |
D89077.1 | - | 2 - 3 | 200 | 116 | |
DA999385.1 | - | 2 - 3 | 357 | 116 | |
DB265860.1 | - | 3 - 4 | 374 | 116 | |
U30473.1 | - | 3 - 4 | 324 | 116 | |
U44403.1 | - | 3 - 4 | 478 | 116 |