Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GTPBP1
Entrez Gene ID
9567
Gene Name
GTP binding protein 1
Gene Aliases
GP-1, GP1, HSPC018
Chromosome Location
Chr.22: 38704561 - 38738265 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004286.4 NP_004277.2 9 - 10 1687 68
XM_011530537.2 XP_011528839.1 9 - 10 2949 68
XM_017029099.1 XP_016884588.1 9 - 10 2941 68
XM_017029100.1 XP_016884589.1 9 - 10 2911 68
XM_017029101.1 XP_016884590.1 10 - 11 1528 68
XM_017029102.1 XP_016884591.1 8 - 9 1344 68
XM_017029103.1 XP_016884592.1 8 - 9 1442 68
GenBank mRNA AI570660.1 - 3 - 4 68
AK315704.1 - 8 - 9 1310 68
BC014075.1 - 9 - 10 1525 68
CR456501.1 - 9 - 10 1510 68
U87964.1 - 9 - 10 1431 68

View Full Product Details