The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001099781.2 | NP_001093251.1 | 5 - 6 | 1177 | 73 |
NM_001099782.2 | NP_001093252.1 | 4 - 5 | 1081 | 73 | |
NM_001302464.1 | NP_001289393.1 | 4 - 5 | 1081 | 73 | |
NM_001302465.1 | NP_001289394.1 | 3 - 4 | 946 | 73 | |
NM_004121.3 | NP_004112.2 | 5 - 6 | 1177 | 73 | |
XM_005261557.2 | XP_005261614.1 | 5 - 6 | 1113 | 73 | |
XM_005261558.2 | XP_005261615.1 | 5 - 6 | 1113 | 73 | |
XM_011530133.1 | XP_011528435.1 | 5 - 6 | 1113 | 73 | |
XM_011530134.1 | XP_011528436.1 | 5 - 6 | 1113 | 73 | |
XM_011530135.1 | XP_011528437.1 | 4 - 5 | 1017 | 73 | |
XM_011530136.1 | XP_011528438.1 | 5 - 6 | 1113 | 73 | |
XM_011530137.2 | XP_011528439.1 | 5 - 6 | 1113 | 73 | |
XM_017028768.1 | XP_016884257.1 | 4 - 5 | 1017 | 73 | |
XM_017028769.1 | XP_016884258.1 | 4 - 5 | 1017 | 73 | |
GenBank mRNA | AK225661.1 | - | 4 - 5 | 1017 | 73 |
AK226019.1 | - | 4 - 5 | 1017 | 73 | |
AK292006.1 | - | 5 - 6 | 959 | 73 | |
AK293683.1 | - | 3 - 4 | 886 | 73 | |
AK297869.1 | - | 6 - 7 | 952 | 73 | |
AK312401.1 | - | 5 - 6 | 1113 | 73 | |
AL117414.1 | - | 4 - 5 | 1022 | 73 | |
BC011362.1 | - | 5 - 6 | 1166 | 73 | |
BC073999.1 | - | 5 - 6 | 1094 | 73 | |
BT009808.1 | - | 5 - 6 | 760 | 73 | |
CT841518.1 | - | 5 - 6 | 918 | 73 | |
M64099.1 | - | 5 - 6 | 1101 | 73 |