The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243225.1 | NP_001230154.1 | 7 - 8 | 860 | 72 |
NM_001282327.1 | NP_001269256.1 | 9 - 10 | 1258 | 72 | |
NM_001282328.1 | NP_001269257.1 | 9 - 10 | 1305 | 72 | |
NM_014303.3 | NP_055118.1 | 7 - 8 | 860 | 72 | |
XM_006724173.1 | XP_006724236.1 | 7 - 8 | 816 | 72 | |
XM_017028678.1 | XP_016884167.1 | 4 - 5 | 2678 | 72 | |
GenBank mRNA | AB451380.1 | - | 7 - 8 | 753 | 72 |
AK096320.1 | - | 9 - 10 | 1277 | 72 | |
AK127185.1 | - | 9 - 10 | 1259 | 72 | |
AK315518.1 | - | 7 - 8 | 827 | 72 | |
BC032489.1 | - | 7 - 8 | 817 | 72 | |
CR456539.1 | - | 7 - 8 | 778 | 72 | |
U78310.1 | - | 7 - 8 | 811 | 72 |