The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014742.3 | NP_055557.2 | 16 - 17 | 1928 | 98 |
XM_005260622.4 | XP_005260679.1 | 16 - 17 | 1980 | 98 | |
XM_017028154.1 | XP_016883643.1 | 15 - 16 | 1625 | 98 | |
XM_017028155.1 | XP_016883644.1 | 15 - 16 | 1896 | 98 | |
XM_017028156.1 | XP_016883645.1 | 16 - 17 | 1676 | 98 | |
XM_017028157.1 | XP_016883646.1 | 16 - 17 | 1933 | 98 | |
XM_017028158.1 | XP_016883647.1 | 15 - 16 | 1587 | 98 | |
XM_017028159.1 | XP_016883648.1 | 16 - 17 | 1733 | 98 | |
XM_017028160.1 | XP_016883649.1 | 16 - 17 | 1645 | 98 | |
XM_017028161.1 | XP_016883650.1 | 15 - 16 | 1597 | 98 | |
GenBank mRNA | AK124411.1 | - | 1 - 2 | 970 | 98 |
AK126534.1 | - | 14 - 15 | 1378 | 98 | |
AK294980.1 | - | 16 - 17 | 1676 | 98 | |
AK296500.1 | - | 16 - 17 | 1585 | 98 | |
AK316293.1 | - | 15 - 16 | 1632 | 98 | |
BC021107.1 | - | 16 - 17 | 1704 | 98 | |
BC022850.2 | - | 15 - 16 | 1686 | 98 | |
D87444.1 | - | 16 - 17 | 1982 | 98 |