The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014742.3 | NP_055557.2 | 5 - 6 | 767 | 80 |
XM_005260622.4 | XP_005260679.1 | 5 - 6 | 819 | 80 | |
XM_017028154.1 | XP_016883643.1 | 5 - 6 | 548 | 80 | |
XM_017028155.1 | XP_016883644.1 | 5 - 6 | 819 | 80 | |
XM_017028156.1 | XP_016883645.1 | 5 - 6 | 515 | 80 | |
XM_017028157.1 | XP_016883646.1 | 5 - 6 | 772 | 80 | |
XM_017028158.1 | XP_016883647.1 | 5 - 6 | 510 | 80 | |
XM_017028159.1 | XP_016883648.1 | 5 - 6 | 600 | 80 | |
XM_017028160.1 | XP_016883649.1 | 5 - 6 | 512 | 80 | |
XM_017028161.1 | XP_016883650.1 | 5 - 6 | 548 | 80 | |
GenBank mRNA | AK294980.1 | - | 5 - 6 | 515 | 80 |
AK296500.1 | - | 5 - 6 | 424 | 80 | |
AK316293.1 | - | 4 - 5 | 471 | 80 | |
BC021107.1 | - | 5 - 6 | 543 | 80 | |
BC022850.2 | - | 4 - 5 | 525 | 80 | |
D87444.1 | - | 5 - 6 | 821 | 80 |