The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014742.3 | NP_055557.2 | 8 - 9 | 1118 | 127 |
XM_005260622.4 | XP_005260679.1 | 8 - 9 | 1170 | 127 | |
XM_017028154.1 | XP_016883643.1 | 8 - 9 | 899 | 127 | |
XM_017028155.1 | XP_016883644.1 | 8 - 9 | 1170 | 127 | |
XM_017028156.1 | XP_016883645.1 | 8 - 9 | 866 | 127 | |
XM_017028157.1 | XP_016883646.1 | 8 - 9 | 1123 | 127 | |
XM_017028158.1 | XP_016883647.1 | 8 - 9 | 861 | 127 | |
XM_017028159.1 | XP_016883648.1 | 8 - 9 | 923 | 127 | |
XM_017028160.1 | XP_016883649.1 | 8 - 9 | 835 | 127 | |
XM_017028161.1 | XP_016883650.1 | 8 - 9 | 871 | 127 | |
GenBank mRNA | AK126534.1 | - | 6 - 7 | 568 | 127 |
AK294980.1 | - | 8 - 9 | 866 | 127 | |
AK296500.1 | - | 8 - 9 | 775 | 127 | |
AK302439.1 | - | 9 - 10 | 973 | 127 | |
AK316293.1 | - | 7 - 8 | 822 | 127 | |
BC021107.1 | - | 8 - 9 | 894 | 127 | |
BC022850.2 | - | 7 - 8 | 876 | 127 | |
D87444.1 | - | 8 - 9 | 1172 | 127 |