Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLSPN
Entrez Gene ID
63967
Gene Name
claspin
Gene Aliases
-
Chromosome Location
Chr.1: 35720465 - 35769985 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190481.1 NP_001177410.1 6 - 7 976 123
NM_022111.3 NP_071394.2 6 - 7 976 123
XM_011541938.2 XP_011540240.1 6 - 7 999 123
XM_011541939.2 XP_011540241.1 6 - 7 993 123
XM_011541940.2 XP_011540242.1 6 - 7 993 123
XM_011541941.2 XP_011540243.1 5 - 6 831 123
GenBank mRNA AF297866.1 - 6 - 7 976 123
AK308193.1 - 6 - 7 1005 123
BC018670.1 - 6 - 7 996 123
BC038991.2 - 6 - 7 1002 123
BC113116.1 - 6 - 7 969 123
BC115025.1 - 6 - 7 969 123
BC115026.1 - 6 - 7 969 123
BC137279.1 - 6 - 7 955 123
BC140789.1 - 6 - 7 955 123

View Full Product Details