Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EIF2B3
Entrez Gene ID
8891
Gene Name
eukaryotic translation initiation factor 2B subunit gamma
Gene Aliases
EIF-2B, EIF2Bgamma
Chromosome Location
Chr.1: 44850522 - 44986722 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166588.2 NP_001160060.1 7 - 8 1011 105
NM_001261418.1 NP_001248347.1 7 - 8 1011 105
NM_020365.4 NP_065098.1 7 - 8 1011 105
XM_017002745.1 XP_016858234.1 7 - 8 899 105
XM_017002746.1 XP_016858235.1 8 - 9 978 105
XM_017002747.1 XP_016858236.1 8 - 9 972 105
GenBank mRNA AF257077.1 - 7 - 8 874 105
AK022399.1 - 7 - 8 873 105
AK024006.1 - 7 - 8 899 105
AK314668.1 - 7 - 8 884 105
AL834288.1 - 7 - 8 876 105
BC018728.1 - 7 - 8 869 105
BC021017.1 - 7 - 8 862 105
CR457285.1 - 3 - 4 265 105

View Full Product Details