The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198665.1 | NP_001185594.1 | 7 - 8 | 859 | 121 |
NM_001301084.1 | NP_001288013.1 | 8 - 9 | 742 | 121 | |
NM_015313.2 | NP_056128.1 | 8 - 9 | 916 | 121 | |
XM_006718805.3 | XP_006718868.1 | 8 - 9 | 1275 | 121 | |
XM_011542720.2 | XP_011541022.1 | 8 - 9 | 857 | 121 | |
XM_017017420.1 | XP_016872909.1 | 7 - 8 | 1492 | 121 | |
XM_017017421.1 | XP_016872910.1 | 8 - 9 | 1132 | 121 | |
XM_017017422.1 | XP_016872911.1 | 8 - 9 | 744 | 121 | |
GenBank mRNA | AB002380.2 | - | 3 - 4 | 278 | 121 |
AF180681.1 | - | 8 - 9 | 588 | 121 | |
AK294803.1 | - | 8 - 9 | 717 | 121 | |
AK304316.1 | - | 8 - 9 | 1140 | 121 | |
BC063117.1 | - | 7 - 8 | 862 | 121 | |
BX647785.1 | - | 8 - 9 | 718 | 121 | |
BX648515.1 | - | 8 - 9 | 723 | 121 |